Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™
GSA_VA

Catalog No. BP243820 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4L
20L
Catalog No. Quantity
BP243820 20L
BP24384 4L
2 options

Catalog No. BP243820

Only null left
Add to Cart

PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4

  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X PBS buffer solution
  • Eliminates the hassle of dilution from stock solutions or weighing and mixing individual components
  • 1X solution contains 0.137M NaCl, 0.0027M KCl, and 0.0119M phosphates
  • Filtered and tested for the absence of DNase, RNase, and protease
  • PBS is used in many biochemical applications to maintain pH
  • Intended for laboratory research use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 0°C
pH 7.3 to 7.5
Physical Form Liquid
Quantity 20L
Packaging Poly Cube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.