Learn More
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™
Shop All Fisherbrand Products

Description
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4
- Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X PBS buffer solution
- Eliminates the hassle of dilution from stock solutions or weighing and mixing individual components
- 1X solution contains 0.137M NaCl, 0.0027M KCl, and 0.0119M phosphates
- Filtered and tested for the absence of DNase, RNase, and protease
- PBS is used in many biochemical applications to maintain pH
- Intended for laboratory research use only
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Melting Point | 0°C |
pH | 7.3 to 7.5 |
Physical Form | Liquid |
Quantity | 4L |
Packaging | Poly Cube |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.