Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™

Catalog No. BP2471100 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle

Catalog No. BP2471-100

Only null left
Add to Cart

TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology

  • Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X TBS buffer solution - simply dilute as needed
  • 10X solution contains 1.37M NaCl, 0.027M KCl and 0.25M tris
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TBS solution with 9 volumes of ultrapure water, such as BP2819
  • Filtered and autoclaved - ready to use stock TBS buffer
  • Tested for the absence of DNase, RNase, and protease
  • TBS buffer is often used in immunodetection techniques such as Western blotting protocols
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris Buffered Saline
Name Note 10X Solution
CAS 7447-40-7, 7647-01-0, 7647-14-5, 77-86-1, 7732-18-5
Boiling Point 100°C
Color Colorless
pH 7.5
Physical Form Liquid
Quantity 100 mL
DNase DNase free
Purity Grade Notes DNase-, RNase- and Protease-Free
Packaging Poly Bottle
Protease Protease free
Synonym TBS
Grade Molecular Biology
Concentration or Composition (by Analyte or Components) 10X solution contains 1.37M Sodium Chloride, 0.027M Potassium Chloride, and 0.25M Tris/Tris-HCl.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.