Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBS Tablets, Phosphate Buffered Saline, Fisher BioReagents™
GSA_VA

Catalog No. BP2944100 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 Tablets
This item is not returnable. View return policy
This item is not returnable. View return policy

Phosphate Buffered Saline Tablets, CAS Number-7647-14-5, 7447-40-7, 100 tablets, Clear, Gray, White, 5/8 in., 600 SEC max., CAUTION!

  • Save time and simplify your buffer preparation step by using Fisher BioReagents PBS tablets
  • Eliminate the hassle of weighing and mixing individual components
  • One tablet dissolved in 200mL water yields buffer containing 0.01M phosphate, 0.0027M KCl, and 0.137M NaCl, pH 7.4 at 25°C
  • Bottle contains 100 tablets
  • Intended for laboratory research use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 7.2 to 7.6
Physical Form Solid
Quantity 100 Tablets
Solution Type Phosphate Buffered Saline
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.