Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Description
Commonly used buffering agent
HEPES is commonly used as a buffering agent.

Specifications
Specifications
Chemical Name or Material | HEPES Buffer |
Absorbance | ≤0.01(0.1M solution) at 260nm, 0.004 max. (0.1M solution) at 280nm |
Name Note | 1M Solution, pH 7.3 |
CAS | 7365-45-9, 7732-18-5 |
Color | Undesignated |
Concentration | 0.95 to 1.05 M |
pH | 7.2 to 7.5 |
Physical Form | Liquid |
Quantity | 1 L |
DNase | DNase free |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction