Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Monobasic Anhydrous (Colorless-to-White Crystals), Fisher BioReagents™

Catalog No. BP329500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle

For tissue culture and molecular biology applications

Sodium Phosphate is commonly used in tissue culture and molecular biology applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
PubChem CID 23672064
ChEBI CHEBI:37585
SMILES [Na+].OP(O)([O-])=O

Specifications

Boiling Point 203.9°C
Melting Point 100°C
Color White
pH 4.1 to 4.5
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99 %
Merck Index 15, 8790
IUPAC Name sodium;dihydrogen phosphate
Formula Weight 119.98
Percent Purity ≥99%
Identification Pass Test
Packaging Glass Bottle
Density 2.04g/cm³
Chemical Name or Material Sodium Phosphate Monobasic Anhydrous
Show More Show Less
Recommended Storage RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.