Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Phosphate Monobasic Monohydrate (Colorless-to-white Crystals), Fisher BioReagents™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Amber Glass
Description
Sodium Phosphate Monobasic Monohydrate (Colorless-to-white Crystals)
Sodium Phosphate is commonly used in tissue culture and molecular biology applications.

Specifications
Specifications
Melting Point | 100°C |
Color | White |
pH | 4.1 to 4.5 |
Physical Form | Solid |
Quantity | 1 kg |
IUPAC Name | sodium phosphoric acid hydrate |
Formula Weight | 137.99 |
Percent Purity | 98.0 to 102.0% |
Identification | Pass Test |
Packaging | Amber Glass |
Show More |
Safety and Handling

Recommended Storage | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction