Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Monobasic Monohydrate (Colorless-to-white Crystals), Fisher BioReagents™

Catalog No. BP330500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Amber Glass

Sodium Phosphate Monobasic Monohydrate (Colorless-to-white Crystals)

Sodium Phosphate is commonly used in tissue culture and molecular biology applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
SMILES O.[Na+].OP(O)(O)=O

Specifications

Melting Point 100°C
Color White
pH 4.1 to 4.5
Physical Form Solid
Quantity 500 g
IUPAC Name sodium;dihydrogen phosphate;hydrate
Formula Weight 137.99
Percent Purity 98.0 to 102.0%
Identification Pass Test
Packaging Amber Glass
Chemical Name or Material Sodium Phosphate Monobasic Monohydrate
Show More Show Less
Recommended Storage RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.