Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper (II) Sulfate Pentahydrate (Crystalline (Blue)), Fisher BioReagents
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Description
Glass Bottle; 500g
Copper Sulfate is suitable for use in tissue culture systems requiring additives.
Specifications
Specifications
| Chemical Name or Material | Copper (II) Sulfate Pentahydrate |
| Melting Point | 110°C |
| Color | Blue |
| CAS | 7758-98-7, 7758-99-8 |
| pH | 3.5 to 4.5 |
| Physical Form | Solid |
| Quantity | 500 g |
| Assay Percent Range | 98.0 to 102.0 % |
| Molecular Formula | CuO4S |
| Merck Index | 15, 2643 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction