Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide (White Pellets), Fisher BioReagents™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Description
Sodium Hydroxide (White Pellets)
Sodium Hydroxide is used to adjust the pH of buffers for molecular biology and cell culture applications.
Specifications
Specifications
| pH | 14 |
| Color | White |
| Boiling Point | 1390°C |
| Melting Point | 318°C |
| UN Number | 1823 |
| Quantity | 2.5 kg |
| Formula Weight | 40 |
| Percent Purity | ≥97.0% |
| Identification | Pass Test |
| Packaging | Poly Bottle |
| Show More |
Safety and Handling
| DOTInformation | DOT Class 8, : Corrosive |
| Recommended Storage | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction