Learn More
PBS, Phosphate Buffered Saline, 10X Solution, Fisher BioReagents™
Shop All Fisherbrand Products
Description
Commonly used buffer
- Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X PBS solution - simply dilute as needed
- 10X solution contains 1.37M NaCl, 0.027M KCl and 0.119M phosphates
- pH of a 1X solution made from this concentrated solution is ∼7.4. To prepare a 1X solution, mix one volume of Fisher BioReagents 10X PBS solution with 9 volumes of ultrapure water, such as BP2819
- Filtered and tested for the absence of DNase, RNase, and protease
- PBS is used in many biochemical applications to maintain pH
- Intended for laboratory research use only
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 500mL |
| Packaging | Poly Bottle |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.