Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents
GSA_VA

Catalog No. BP82005 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Poly Bottle
Poly Pail

Sodium Dodecyl Sulfate is the most commonly used detergent in protein purification and electrophoresis

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Absorbance ≤0.10 (0.1% Solution) at 260nm
Color White
Physical Form Solid
Melting Point 204°C
pH 8.5 to 10
Quantity 5 kg
Identification Pass test
Packaging Poly Pail
Assay Percent Range >99%
IUPAC Name sodium dodecyl sulfate
Percent Purity ≥98.0% (C12 alcohol)
Grade Molecular Biology
Chemical Name or Material Sodium Dodecyl Sulfate (SDS), Micropellets
Show More Show Less
Recommended Storage Store at room temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.