Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentamycin Sulfate (White Powder), Fisher BioReagents

Catalog No. BP9181 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Catalog No. BP918-1

Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

For selecting transformed cells containing gentamycin resistance gene

Gentamycin Sulfate inhibits protein synthesis, preventing microbial contamination in tissue culture applications. It is also used to confer antibiotic resistance/sensitivity in molecular biology procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-41-0
Molecular Formula C21H43N5O7·H2SO4
Synonym Gentiomycin C, Garamycin

Specifications

Type Gentamicin Sulfate
Physical Form Solid
pH 3.5 to 5.5
Melting Point 218°C
Ignition Residue < 1.0%
Packaging Amber Glass
Potency > 590μg/mg
Specific Rotation Inclusive between 107° (+ or -)
Color White
Quantity 1 g
Merck Index 15, 4427
Formula Weight 575.67
Chemical Name or Material Gentamycin Sulfate
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
May impair fertility. May cause skin, eye, and respiratory tract irritation. May cause allergic respiratory and skin reaction. May cause adverse kidney effects. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Use personal protective equipment. Do not breathe vapors/dust. Do not ingest. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage 4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.