Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform (Ethanol as Preservative/Certified ACS), Fisher Chemical™

Catalog No. C2984 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Aluminum Can
Glass Bottle
Steel Drum
Steel Pail

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3, 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Color Colorless
Decomposition Information 290°C
Density 1.480g/cm³
Boiling Point 61°C
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range ≥99.8 %
Vapor Pressure 213mbar at 20°C
Viscosity 0.56 mPa/s at 20°C
Water 0.02% max.
Formula Weight 119.38
Percent Purity ≥99.8%
Grade Certified ACS Reagent
Concentration or Composition (by Analyte or Components) 0.75% Ethanol
Chemical Name or Material Chloroform, Approx. 0.75% ethanol as preservative
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:2
Flammability:1
Instability:0
DOTInformation DOT Class 6.1, : Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.