Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane (Spectranalyzed™), Fisher Chemical™

Catalog No. C5551 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass

For use in the ultraviolet and infrared ranges

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Absorbance Pass Test
Color Colorless
Density 0.770g/cm³
Boiling Point 81°C
Quantity 1 L
Flash Point -18°C (64.4°F)
Identification Pass Test
Infrared Spectrum Pass Test
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range ≥99.0 %
Linear Formula CH2(CH2)4CH2
Vapor Pressure 104mbar at 20°C
Viscosity 0.94 mPa/s at 20°C
UN Number 1145
Water 0.02% max.
Formula Weight 84.16
Percent Purity ≥99.0%
Grade Also meets ACS specifications
Chemical Name or Material Cyclohexane
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.