Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloroform (Ethanol as Preservative, Spectranalyzed™), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Specifications
Specifications
| Melting Point | -63°C |
| Absorbance | Pass Test |
| Color | Colorless |
| Decomposition Information | 290°C |
| Density | 1.480g/cm³ |
| Boiling Point | 61°C |
| Quantity | 4 L |
| Identification | Pass Test |
| Infrared Spectrum | Conforms to Ref |
| Packaging | Glass Bottle |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. NFPA Health:2 Flammability:1 Instability:0 |
| DOTInformation | DOT Class 6.1, : Poison |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction