Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride (Desiccant, 20 mesh and Finer) ACS Grade, Fisher Chemical™

Catalog No. C77212 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Color Beige
Boiling Point 1600°C
Melting Point 782°C
Physical Form Solid
pH 8 to 10
Material Calcium Chloride
Quantity 2.5 kg
Identification Pass Test
Assay Percent Range ≥96 %
Mesh Size 20 mesh and finer
Packaging Poly Bottle
Formula Weight 110.99
Percent Purity ≥96%
Grade ACS Grade
Chemical Name or Material Calcium Chloride, Desiccant
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.