Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cal-Ex™ Decalcifier, Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
10 L
Packaging:
Poly Bottle
PolyPac™
Specifications
Specifications
Physical Form | Liquid |
Chemical Name or Material | Cal-Ex™ Decalcifier |
Boiling Point | 100°C |
CAS | 6381-92-6, 7647-01-0, 7732-18-5 |
Packaging | PolyPac™ |
pH | 2.0 |
Quantity | 10 L |
Assay Percent Range | ≥98.5 % |
MDL Number | MFCD00211743 |
Concentration or Composition (by Analyte or Components) | 0.0027 to 0.0038M (1.35 to 1.40N) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction