Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N,N-Dimethylformamide (Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
HDPE Drum
Poly Pail
Specifications
Specifications
| pH | 6 to 8 |
| Melting Point | -61°C |
| Color | Colorless |
| Decomposition Information | >350°C |
| Density | 0.945g/cm³ |
| Boiling Point | 153°C |
| Quantity | 1 L |
| Flash Point | 58°C (136.4°F) |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. May cause methemoglobinemia. Irritating to eyes and skin. Lachrymator (substance which increases the flow of tears). May cause central nervous system effects. May cause adverse liver effects. May cause adverse kidney effects. May cause harm to the unborn child. Use personal protective equipment. Keep away from open flames |
| DOTInformation | DOT Class 3, : Flammable Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction