Learn More
Specifications
Specifications
| pH | 6 to 8 |
| Melting Point | -61°C |
| Absorbance | Pass Test |
| Color | Colorless |
| Decomposition Information | >350°C |
| Density | 0.945g/cm³ |
| Boiling Point | 153°C |
| Quantity | 4 L |
| Identification | Pass Test |
| Infrared Spectrum | Conforms to reference |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. May cause methemoglobinemia. Irritating to eyes and skin. Lachrymator (substance which increases the flow of tears). May cause central nervous system effects. May cause adverse liver effects. May cause adverse kidney effects. May cause harm to the unborn child. Use personal protective equipment. Keep away from open flames |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.