Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMSO, Dimethyl Sulfoxide, GC Headspace Grade, Thermo Scientific™

Catalog No. D1391 Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
Packaging:
Glass Bottle

DMSO processed to high purity for accurate and repeatable determination of trace levels of residual solvents.

  • Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
  • 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
  • This product is UV Absorbance Tested to ensure low organic contamination
  • Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Absorbance 0.40 AU Maximum (270nm), 0.18 AU Maximum (290nm), 0.06 AU Maximum (310nm), 0.02 AU Maximum (330nm), 0.01 AU Maximum (350 to 400nm)
Color Colorless
Quantity 1 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.01 % Maximum
Titratable Acid 0.001 mEq/g Maximum
Water 0.05% Maximum
Formula Weight 78.13
Percent Purity ≥99.9%
Grade GC Headspace
Chemical Name or Material DMSO, Dimethyl Sulfoxide
Show More Show Less
Recommended Storage Room temperature, packaged under inert atmosphere.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.