Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride (Stabilized/Certified ACS), Fisher Chemical™

Catalog No. D3720 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass 
Steel Drum
Steel Pail

Catalog No. D37-20

Only null left
Add to Cart

Stabilized with preservative. Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2, 67-56-1, 110-83-8, 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Density 1.33g/cm³
Boiling Point 39°C
Quantity 20 L
Identification Pass Test
Packaging Steel Pail
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range ≥99.5 %
Titratable Acid 0.0003mEq/g max.
Vapor Pressure 350mbar at 20°C
Water 0.02% max.
Formula Weight 84.93
Percent Purity ≥99.5%
Grade Certified ACS Reagent
Concentration or Composition (by Analyte or Components) Methyl alcohol, Cyclohexene, 2-Methyl-2-butene
Chemical Name or Material Methylene Chloride, Stabilized
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
DOTInformation DOT Class 6.1, : Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.