Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylene Chloride (Stabilized/Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass
Steel Drum
Steel Pail
Specifications
Specifications
Melting Point | -97°C |
Color | Colorless |
Density | 1.33g/cm³ |
Boiling Point | 39°C |
Quantity | 4 L |
Identification | Pass Test |
Packaging | Glass Bottle |
Physical Form | Liquid |
Assay Percent Range | ≥99.5 % |
Residue after Evaporation | 0.002% max. |
Show More |
Safety and Handling


Hazard Statement | WARNING! |
Precautionary Statement | Emergency Overview Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. NFPA Health:2 Flammability:1 Instability:0 |
DOTInformation | DOT Class 6.1, : Poison |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction