Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Ethyl Ether Anhydrous (Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                500 mL
                            
                        
                            
                                1 L
                            
                        
                            
                                4 L
                            
                        
                            
                                20 L
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Aluminum Can
                            
                        
                            
                                Steel Pail
                            
                        
                    Description
Stabilized with BHT as preservative. Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
            Specifications
Specifications
| Melting Point | -116°C | 
| Color | Colorless | 
| Density | 0.714g/cm³ | 
| Boiling Point | 34.6°C | 
| Quantity | 4 L | 
| Flash Point | -45°C (-111°F) | 
| Identification | Pass Test | 
| Packaging | Aluminum Can | 
| Physical Form | Liquid | 
| Residue after Evaporation | 0.001% max. | 
| Show More | 
Safety and Handling
                        
                        
                            
                        
                        | Hazard Statement | DANGER! | 
| Precautionary Statement | Emergency Overview  Extremely flammable liquid and vapor. Vapor may cause flash fire. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Hygroscopic. Use personal protective equipment. Keep away from open flames  | 
                                
| DOTInformation | DOT Class 3, : Flammable Liquid | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction