Learn More
Description
Stabilized with BHT as preservative. Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
Specifications
Specifications
| Melting Point | -116°C |
| Color | Colorless |
| Density | 0.714g/cm³ |
| Quantity | 500 mL |
| Flash Point | -45°C (-111°F) |
| Identification | Pass Test |
| Packaging | Aluminum Can |
| Physical Form | Liquid |
| Assay Percent Range | ≥99 % |
| Residue after Evaporation | 0.001% max. |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Extremely flammable liquid and vapor. Vapor may cause flash fire. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Hygroscopic. Use personal protective equipment. Keep away from open flames |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.