Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Petroleum Ether (Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Aluminum Can
Glass Bottle
Steel Drum
Steel Pail
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Color | Colorless |
Acidity | Pass Test |
Boiling Point | 38°C |
Quantity | 20 L |
Identification | Pass Test |
Packaging | Steel Pail |
Physical Form | Liquid |
Residue after Evaporation | 0.001% max. |
Specific Gravity | 0.630 to 0.660 (at 25°C) |
Grade | Certified ACS Reagent |
Show More |
Safety and Handling


Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Flammable liquid and vapor. May cause heritable genetic damage. May cause eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air. NFPA Health:1 Flammability:3 Instability:0 |
DOTInformation | DOT Class 3, : Flammable Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction