Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Ether (Laboratory), Fat Extraction, Fisher Chemical™

Catalog No. E49220 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
20 L
Packaging:
Aluminum Can
Steel Pail

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Color Colorless
Density 0.714g/cm³
Boiling Point 34.6°C
Quantity 20 L
Flash Point -45°C (113°F)
Identification Pass Test
Packaging Steel Pail
Physical Form Liquid
Linear Formula C2H5OC2H5
Vapor Pressure 587mbar at 20°C
Viscosity 0.2448 cP at 20°C
Formula Weight 74.12
Grade Suitable for fat extraction
Chemical Name or Material Ethyl Ether
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.