Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde (USP), Fisher Chemical™
GSA_VA

Catalog No. F77200 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Poly Bottle
Poly Pail
PolyPac™
Steel Drum

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0, 67-56-1, 7732-18-5
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Acidity Pass Test
Melting Point 0°C
Color Colorless
Boiling Point 101°C
Assay Percent Range 36.5 to 38.0 %
Packaging Steel Drum
Linear Formula HCHO
Quantity 200 L
Formula Weight 30.03
Percent Purity ≥34.5%
Grade USP
Concentration or Composition (by Analyte or Components) 40% by Volume
Physical Form Liquid
Chemical Name or Material Formaldehyde, 40% by Volume
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.

NFPA

Health:3
Flammability:2
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.