Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formaldehyde, 37% by Weight (With Preservative/Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
4 L
200 L
Packaging:
HDPE Bottle
Poly Bottle
Poly Drum
Specifications
Specifications
| Melting Point | 0°C |
| Color | Colorless |
| Boiling Point | 101°C |
| Ignition Residue | 0.005% max. |
| Assay Percent Range | 36.5 to 38.0 % |
| Packaging | Poly Bottle |
| Linear Formula | HCHO |
| Quantity | 4 L |
| Formula Weight | 30.03 |
| Percent Purity | 36.5 to 38.0% |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. NFPA Health:3 Flammability:2 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction