Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Hydrogen Peroxide, Fisher Chemical™
Shop All Fisherbrand Products
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                500 mL
                            
                        
                            
                                4 L
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Glass Bottle
                            
                        
                            
                                Poly Bottle
                            
                        
                    Specifications
Specifications
| Color | Undesignated | 
| Acidity | Pass Test | 
| Boiling Point | 100°C | 
| Melting Point | 0°C | 
| Assay Percent Range | 3% w/v | 
| Density | 1.00g/cm³ | 
| Formula Weight | 34.01 | 
| Percent Purity | 3.0 to 3.5% | 
| Grade | USP | 
| Concentration or Composition (by Analyte or Components) | 3% w/v | 
| Show More | 
Safety and Handling
                        
                        | Hazard Statement | CAUTION! | 
| Precautionary Statement | Emergency Overview  May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. Note: Products are tested to meet the analytical requirements of the noted grade (USP, Ph. EUR or otherwise). Product test results are available on the Certificate of Analysis or upon request for the specific product. Products are for research use or further manufacturing. Products are not for direct administration to humans or animals. It is the responsibility of the final formulator and end-user to qualify, test and/or validate each product for its intended use as an ingredient in the end product and in accordance with applicable regulations (including without limitation WHO-recommended Hand Rub Formulations (Hand Sanitizers) and/or FDA Policy for Preparation of Certain Alcohol-based Hand Sanitizer Products, as updated). Thermo Fisher Scientific expressly disclaims all non-specified warranties, expressed or implied, including the implied warranties of merchantability and fitness for a particular use. NFPA Health:1 Flammability:1 Instability:0  | 
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction