Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
500 mL
4 L
30 gal.
Packaging:
Poly Bottle
Poly Drum
Description
Hydrogen Peroxide, 30% (Certified ACS)
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 108°C |
| Melting Point | -33°C |
| Assay Percent Range | 29.0 to 32.0 % |
| Density | 1.110g/cm³ |
| Formula Weight | 34 |
| Percent Purity | 29.0 to 32.0% |
| Grade | ACS |
| Identification | Pass test |
| Packaging | Poly Drum |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: DANGER! |
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Causes severe eye irritation and possible burns. Irritating to skin. Harmful if swallowed. NFPA Health:3 Flammability:0 Instability:1 |
| DOTInformation | DOT Class 5.1, : Oxidizer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction