Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™
GSA_VA

Catalog No. H325500 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
500 mL
4 L
30 gal.
Packaging:
Poly Bottle
Poly Drum

Hydrogen Peroxide, 30% (Certified ACS)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1, 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Boiling Point 108°C
Melting Point -33°C
Assay Percent Range 29.0 to 32.0 %
Density 1.110g/cm³
Formula Weight 34
Percent Purity 29.0 to 32.0%
Grade ACS
Identification Pass test
Packaging Poly Bottle
Residue after Evaporation 0.002 % max.
Vapor Pressure 23mmHg at 30°C
Decomposition Information Decomposes information: >125°C
pH 3.3
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide, 30%
Show More Show Less
Hazard Category Signal Word: DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes severe eye irritation and possible burns. Irritating to skin. Harmful if swallowed.
NFPA
Health:3 Flammability:0 Instability:1
DOTInformation DOT Class 5.1, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.