Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hyflo™ SuperCel™ Filter Aid, Fisher Chemical
Shop All Fisherbrand Products
Click to view available options
Quantity:
3 kg
Specifications
Specifications
Physical Form | Solid |
Chemical Name or Material | Hyflo™ SuperCel™ Filter Aid |
Melting Point | 1710°C |
Quantity | 3 kg |
Color | Gray |
pH | 5 to 10 |
Boiling Point | 2230°C |
CAS | 68855-54-9, 14464-46-1, 14808-60-7 |
Packaging | Poly Pail |
Molecular Formula | SiO2 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction