Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water with 0.1% Trifluoroacetic Acid (v/v), Optima™ LC/MS Grade, Thermo Scientific™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
Description
- Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
- Pre-mixed for ease-of-use
- Low metal ion content to ensure minimal metal contamination
- TFA is a common mobile phase additive that can function as an ion pair reagent
Specifications
Specifications
| Absorbance | Report AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm |
| Assay Percent Range | 99.0 to 103.0 % |
| Molecular Formula | C2HF3O2 |
| MDL Number | MFCD00004169 |
| Merck Index | 15, 9847 |
| Synonym | trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova |
| InChI Key | DTQVDTLACAAQTR-UHFFFAOYSA-N |
| SMILES | OC(=O)C(F)(F)F |
| IUPAC Name | trifluoroacetic acid |
| Molecular Weight (g/mol) | 114.02 |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:1 Flammability:0 Instability:0 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction