Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium Oxide (Heavy Powder/USP/FCC), Fisher Chemical™
Shop All Fisherbrand Products

Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
Poly Pail
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Boiling Point | 3600°C |
| Melting Point | 2800°C |
| Color | White |
| pH | 10.3 |
| Physical Form | Powder/Solid |
| Quantity | 3 kg |
| Formula Weight | 40.3 |
| Percent Purity | 96.0 to 100.5% (USP) |
| Grade | FCC/USP |
| Identification | Pass Test (FCC), Pass Test (FCC/USP), Pass Test (USP) |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: CAUTION! |
| Precautionary Statement | Emergency Overview May cause eye, skin and respiratory tract irritation. NFPA Health: 1 Flammability: 0 Instability: 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction