Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChromaCare™ LC-MS Biologics Flush Solution

Catalog No. MB1241 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Borosilicate Glass Bottle

Catalog No. MB1241

Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.

Flush solution for cleaning blood, plasma, and urine samples from the chromatograph.

  • Suitable for flushing difficult to clean samples such as blood, plasma, and urine after vitamin D, hormone or drugs of abuse testing
  • Save time by using off-the-shelf blend — premade for convenience
  • Helps to reduce sample carryover
  • Packaged in 2.5L amber glass bottle
TRUSTED_SUSTAINABILITY

Specifications

Absorbance ≤1.0 AU at 210nm, ≤1.0 AU at 220nm
Chemical Name or Material 2-Propanol (45%), Acetonitrile (45%), and Acetone (10%) Solution
pH 6 to 7
Name Note Mobile-phase solvent blend
Color Colorless
Boiling Point 75°C
CAS 67-64-1, 75-05-8
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Percent Purity Pass Test (By GC-FID)
Grade LC-MS
Concentration or Composition (by Analyte or Components) 45% 2-Propanol: 45% Acetonitrile: 10% Acetone
Packaging Borosilicate Glass Bottle
Density 0.787 g/cm3
Quantity 1 L
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.