Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate Anhydrous (Powder/Certified ACS), Fisher Chemical™

Catalog No. S2633 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Catalog No. S263-3

Only null left
Add to Cart

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Boiling Point 1600°C
Melting Point 854°C
Color White
pH 11.3
Physical Form Solid
Quantity 3 kg
Assay Percent Range ≥99.5%
Formula Weight 105.99
Percent Purity ≥99.5%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Density 2.53g/cm³
Assay (dried basis)≥99.5%
Chemical Name or Material Sodium Carbonate Anhydrous
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Use personal protective equipment. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:0
Instability:1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.