Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. S343500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle

Catalog No. S343-500

Only null left
Add to Cart

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Boiling Point 380°C
Melting Point 306°C
Color White
pH 5.5 to 8
Physical Form Crystalline Solid
Quantity 500 g
Assay Percent Range ≥99 %
UN Number 1498
Formula Weight 84.99
Percent Purity ≥99%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Sodium Nitrate
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Irritating to eyes, respiratory system and skin. May cause cancer. Wear personal protective equipment. Keep away from clothing and other combustible materials. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:1
Instability:3
DOTInformation DOT Class 5.1, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.