Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide (Pellets/NF/FCC/EP/BP/JP), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
Poly Drum
Poly Pail
Specifications
Specifications
| pH | 14 |
| Color | White |
| Boiling Point | 1390°C |
| Melting Point | 318°C |
| Density | 2.13g/cm³ |
| Quantity | 50 kg |
| Formula Weight | 40 |
| Percent Purity | 95.0 to 100.5% (NF) |
| Grade | BP/EP/FCC/JP/NF |
| Identification | Pass Test (BP), Pass Test (BP/EP/FCC/JP/NF), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (NF) |
| Show More |
Safety and Handling
| DOTInformation | DOT Class 8, : Corrosive |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction