Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Thiosulfate Anhydrous (Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
Specifications
Specifications
| Melting Point | 48°C |
| Color | White |
| pH | 6.0 to 8.5 |
| Physical Form | Solid |
| Quantity | 3 kg |
| Assay Percent Range | ≥98 % |
| Formula Weight | 158.11 |
| Percent Purity | ≥98% |
| Grade | Certified |
| Identification | Pass Test |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction