Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride (colorless to white crystalline granules/USP/FCC/EP/BP), Fisher Chemical™
GSA_VA

Catalog No. S641P350LB Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
350 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Drum

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 350 lb.
Formula Weight 58.44
Percent Purity 99.0 to 100.5% (BP)
Grade BP/EP/FCC/USP
Identification Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC/EP/BP)
Loss on Drying 0.5% max. (BP), 0.5% max. (EP), 0.5% max. (FCC), 0.5% max. (USP), 0.5% max. (USP/FCC/EP/BP)
Packaging Poly Drum
Vapor Pressure 1mmHg at 865°C
Density 2.165g/cm³
Chemical Name or Material Sodium Chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.