Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel Desiccant (6-14 Mesh, Certified ACS), Fisher Chemical™

Catalog No. S68410 Shop All Fisherbrand Products
Click to view available options
Quantity:
2.5 kg
10 kg
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
MDL Number MFCD00011232
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Boiling Point 2230°C
Color White
Melting Point 1710°C
Physical Form Solid
pH 2.3 to 7.4
Density 2.1g/cm³
Material Silica Gel
Quantity 10 kg
Identification Pass Test
Mesh Size 6 to 14 Mesh
Packaging Poly Pail
Suitability Pass Test
Grade Certified ACS
Chemical Name or Material Silica Gel Desiccant, 6-14 Mesh
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.