Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution, pH 6.00 (Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Poly Bottle
PolyPac™
Specifications
Specifications
| Chemical Name or Material | Buffer Solution, pH 6.00 |
| Melting Point | 0°C |
| CAS | 7778-77-0, 7732-18-5, 1310-73-2 |
| Boiling Point | 100°C |
| Color | Undesignated |
| pH | 6.00 |
| Physical Form | Liquid |
| Quantity | 20 L |
| Identification | Pass Test |
| Packaging | PolyPac™ |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction