Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution, pH 2.00 (Certified), Fisher Chemical™
Shop All Fisherbrand Products

missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
1 L
20 L
Packaging:
Poly Bottle
PolyPac™
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Chemical Name or Material | Buffer Solution, pH 2.00 |
Melting Point | 0°C |
CAS | 7447-40-7, 7647-01-0, 50-00-0, 67-56-1, 7732-18-5 |
Boiling Point | 100°C |
Color | Undesignated |
Density | 1.0g/cm³ |
pH | 2.00 |
Physical Form | Liquid |
Quantity | 500 mL |
Identification | In Pass Test |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction