Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
Specifications
Specifications
Packaging | Poly Bottle |
Vapor Pressure | 14mmHg at 20°C |
Density | 1.0g/cm³ |
Physical Form | Liquid |
Concentration | 990 to 1010 ppm |
Color | Orange |
pH | 7 |
Melting Point | 0°C |
Boiling Point | 100°C |
Quantity | 500 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction