Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formalin, Buffered, 10% (Phosphate Buffer/Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Poly Bottle
Poly Drum
PolyPac™
Specifications
Specifications
| Physical Form | Liquid |
| Chemical Name or Material | Formalin, Buffered, 10% |
| Melting Point | 0°C |
| Name Note | Phosphate Buffer |
| Boiling Point | 93.9°C |
| CAS | 50-00-0, 67-56-1, 7558-79-4, 10049-21-5, 7732-18-5 |
| Density | 1.10g/cm³ |
| Packaging | Poly Drum |
| pH | 6.9 to 7.1 |
| Quantity | 200 L |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction