Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride Solution, 0.025% w/v (APHA), Fisher Chemical

Catalog No. SF971 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Catalog No. Quantity Packaging
SF97-1 1 L Glass Bottle
1 options

Catalog No. SF97-1

Only null left
Add to Cart

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7705-08-0 , 7732-18-5
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
Synonym Iron (III) Chloride Solution, 0.025% w/v
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 100°C
Melting Point 0°C
Color Yellow
pH 2.54
Physical Form Liquid
Quantity 1 L
UN Number 1759
Grade APHA
Concentration or Composition (by Analyte or Components) 0.025% w/v
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 760mmHg at 100°C
Density 1g/cm³
Chemical Name or Material Ferric Chloride Solution, 0.025% (w/v)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.