Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formalin Solution, 10% (Histological), Fisher Chemical™

Catalog No. SF9820 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Bottle
PolyPac™

Chemical Identifiers

CAS 50-00-0, 67-56-1, 7732-18-5
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Physical Form Liquid
Melting Point 0°C
Boiling Point 100°C
Packaging PolyPac™
Color Colorless
Quantity 20 L
Percent Purity 3.5 to 4.5%
Grade Histological
Concentration or Composition (by Analyte or Components) 10% w/v
Chemical Name or Material Formalin Solution, 10%
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Cancer hazard. Cannot be made non-poisonous. Harmful if absorbed through skin or if inhaled. Vapor harmful. Irritating to eyes, respiratory system and skin. May cause an allergic skin reaction. Possible risks of irreversible effects. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:1
Instability:0
DOTInformation DOT Class 9, : Miscellaneous
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.