Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Solution, Wijs, Fisher Chemical™

Catalog No. SI1064 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass

For determination of iodine absorption number in fats. Approximately 0.22N solution in glacial acetic acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2, 64-19-7, 7782-50-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Amber
Boiling Point 118°C
Melting Point 17°C
UN Number 2920
Concentration 0.215 to 0.225
Density 1.06g/cm³
Identification Pass test
Packaging Amber Glass
Vapor Pressure 11mmHg at 20°C
pH 2.0
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Iodine Solution, Wijs
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:2
Instability:0
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.