Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein Solution, Alcoholic, 1.0%, Fisher Chemical™

Catalog No. SP621 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Catalog No. SP62-1

Only null left
Add to Cart
This item is not returnable. View return policy

Volumetric indicator

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8, 67-63-0
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point -89°C
Boiling Point 83°C
Concentration 0.7 to 1.3%
Concentration or Composition (by Analyte or Components) 1% w/v
Packaging Glass Bottle
Vapor Pressure 40mmHg
Visual Transition Interval Pass Test
Density 0.7855g/cm³
Color Colorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Phenolphthalein Solution, Alcoholic 1.0%
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.